Benzyl phenyl ether, 98%, Thermo Scientific Chemicals
Benzyl phenyl ether, 98%, Thermo Scientific Chemicals
Benzyl phenyl ether, 98%, Thermo Scientific Chemicals
Benzyl phenyl ether, 98%, Thermo Scientific Chemicals
Acros Organics
Benzyl phenyl ether, 98%, Thermo Scientific Chemicals
CAS: 946-80-5 | C13H12O | 184.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Catalog number ACR310260250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS126-73-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Refractive index1.4230 to 1.4250 (20°C, 589 nm)
Infrared spectrumConforms
GC>=99.0 %
Color scale=<50 APHA