D-Glucuronic acid, 98%, Thermo Scientific Chemicals
D-Glucuronic acid, 98%, Thermo Scientific Chemicals
D-Glucuronic acid, 98%, Thermo Scientific Chemicals
D-Glucuronic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
D-Glucuronic acid, 98%, Thermo Scientific Chemicals
CAS: 6556-12-3 | C6H10O7 | 194.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
Packaging:
Plastic bottle
Catalog number ACR309880100
Price (MYR)
854.00
Add to cart
Quantity:
10 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
854.00
Add to cart
Chemical Identifiers
CAS1921-70-6
IUPAC Name2,6,10,14-tetramethylpentadecane
Molecular FormulaC19H40
InChI KeyXOJVVFBFDXDTEG-UHFFFAOYNA-N
SMILESCC(C)CCCC(C)CCCC(C)CCCC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=94.0 %
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid