4-Methoxyphenylboronic acid, 97%, Thermo Scientific Chemicals
4-Methoxyphenylboronic acid, 97%, Thermo Scientific Chemicals
4-Methoxyphenylboronic acid, 97%, Thermo Scientific Chemicals
4-Methoxyphenylboronic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Methoxyphenylboronic acid, 97%, Thermo Scientific Chemicals
CAS: 5720-07-0 | C7H9BO3 | 151.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number ACR309480050
Price (MYR)
730.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
730.00
Add to cart
Chemical Identifiers
CAS56-92-8
IUPAC Namedihydrogen 2-(1H-imidazol-5-yl)ethan-1-amine dichloride
Molecular FormulaC5H11Cl2N3
InChI KeyPPZMYIBUHIPZOS-UHFFFAOYSA-N
SMILES[H+].[H+].[Cl-].[Cl-].NCCC1=CN=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<0.5 % (105°C)
Sulfated ash=<0.1 %
Appearance (Color)White
Appearance (Form)Crystalline powder