Nonafluoropentanoic acid, 97%, Thermo Scientific Chemicals
Nonafluoropentanoic acid, 97%, Thermo Scientific Chemicals
Nonafluoropentanoic acid, 97%, Thermo Scientific Chemicals
Nonafluoropentanoic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Nonafluoropentanoic acid, 97%, Thermo Scientific Chemicals
CAS: 2706-90-3 | C5HF9O2 | 264.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
5 mL
Packaging:
Glass bottle
Catalog number ACR309290250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2745-49-5
IUPAC Name1,4-dichloro-2-(chloromethyl)benzene
Molecular FormulaC7H5Cl3
InChI KeyOMZINLIPPVNUOG-UHFFFAOYSA-N
SMILESClCC1=CC(Cl)=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥96.0%
Refractive Index1.5725-1.5775 @ 20°C