Nonafluoropentanoic acid, 97%, Thermo Scientific Chemicals
Nonafluoropentanoic acid, 97%, Thermo Scientific Chemicals
Nonafluoropentanoic acid, 97%, Thermo Scientific Chemicals
Nonafluoropentanoic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Nonafluoropentanoic acid, 97%, Thermo Scientific Chemicals
CAS: 2706-90-3 | C5HF9O2 | 264.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Catalog number ACR309290050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS543-20-4
IUPAC Namebutanedioyl dichloride
Molecular FormulaC4H4Cl2O2
InChI KeyIRXBNHGNHKNOJI-UHFFFAOYSA-N
SMILESClC(=O)CCC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration RCOCl>=94 %
Appearance (Form)Liquid
Refractive index1.4670 to 1.4720 (20°C, 589 nm)
Appearance (Color)Clear colorless to light brown