4'-tert-Butylacetophenone, 98%, Thermo Scientific Chemicals
4'-tert-Butylacetophenone, 98%, Thermo Scientific Chemicals
4'-tert-Butylacetophenone, 98%, Thermo Scientific Chemicals
4'-tert-Butylacetophenone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4'-tert-Butylacetophenone, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number ACR308740010
Price (MYR)
489.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
489.00
Add to cart
Specifications
Chemical Name or Material4'-tert-Butylacetophenone
Name Note98%
CAS943-27-1
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning