1-Bromo-3-methyl-2-butene, 96%, Thermo Scientific Chemicals
1-Bromo-3-methyl-2-butene, 96%, Thermo Scientific Chemicals
1-Bromo-3-methyl-2-butene, 96%, Thermo Scientific Chemicals
1-Bromo-3-methyl-2-butene, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Bromo-3-methyl-2-butene, 96%, Thermo Scientific Chemicals
CAS: 870-63-3 | C5H9Br | 149.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Catalog number ACR307410100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS2926-30-9
IUPAC Namesodium trifluoromethanesulfonate
Molecular FormulaCF3NaO3S
InChI KeyXGPOMXSYOKFBHS-UHFFFAOYSA-M
SMILES[Na+].[O-]S(=O)(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Powder
Infrared spectrumConforms
Assay>=97.5 % (ex Na)