Bromonitromethane, 90%, Tech., Thermo Scientific Chemicals
Bromonitromethane, 90%, Tech., Thermo Scientific Chemicals
Bromonitromethane, 90%, Tech., Thermo Scientific Chemicals
Bromonitromethane, 90%, Tech., Thermo Scientific Chemicals
Acros Organics
Bromonitromethane, 90%, Tech., Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Packaging:
Glass bottle
Catalog number ACR307050010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialBromonitromethane
Name Notetech., 90%
CAS563-70-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May intensify fire; oxidizer.