2,6-Dimethoxybenzaldehyde, 99%
2,6-Dimethoxybenzaldehyde, 99%
2,6-Dimethoxybenzaldehyde, 99%
2,6-Dimethoxybenzaldehyde, 99%
Thermo Scientific Chemicals
2,6-Dimethoxybenzaldehyde, 99%
CAS: 3392-97-0 | C9H10O3 | 166.18 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR3057900101 gGlass bottle
Catalog number ACR305790010
Price (MYR)
598.00
EA
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
598.00
EA
Add to cart
Chemical Identifiers
CAS127-08-2
IUPAC Namepotassium acetate
Molecular FormulaC2H3KO2
InChI KeySCVFZCLFOSHCOH-UHFFFAOYSA-M
SMILES[K+].CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Insoluble material0.005%
Impurity contentCa 0.005%
Assay (unspecified)99% C₂H₃KO₂ min
Heavy Metals(ICP-OES) 5ppm
Impurity contentCl 0.003%