4-Fluoro-3-nitrobenzoic acid, 98%, Thermo Scientific Chemicals
4-Fluoro-3-nitrobenzoic acid, 98%, Thermo Scientific Chemicals
4-Fluoro-3-nitrobenzoic acid, 98%, Thermo Scientific Chemicals
4-Fluoro-3-nitrobenzoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Fluoro-3-nitrobenzoic acid, 98%, Thermo Scientific Chemicals
CAS: 453-71-4 | C7H4FNO4 | 185.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number ACR305060250
Price (MYR)
1,409.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,409.00
Add to cart
Specifications
Chemical Name or Material4-Fluoro-3-nitrobenzoic acid
Name Note98%
CAS453-71-4
CAS Min %97.5
CAS Max %100.0