4-Methyl-1,3-dioxane, 99%, Thermo Scientific Chemicals
4-Methyl-1,3-dioxane, 99%, Thermo Scientific Chemicals
4-Methyl-1,3-dioxane, 99%, Thermo Scientific Chemicals
4-Methyl-1,3-dioxane, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Methyl-1,3-dioxane, 99%, Thermo Scientific Chemicals
CAS: 1120-97-4 | C5H10O2 | 102.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number ACR305000050
Price (MYR)
416.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
416.00
Add to cart
Chemical Identifiers
CAS7440-05-3
IUPAC Namepalladium
Molecular FormulaPd
InChI KeyKDLHZDBZIXYQEI-UHFFFAOYSA-N
SMILES[Pd]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.05% max.