Harmine, 98%, Thermo Scientific Chemicals
Harmine, 98%, Thermo Scientific Chemicals
Harmine, 98%, Thermo Scientific Chemicals
Harmine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Harmine, 98%, Thermo Scientific Chemicals
CAS: 442-51-3 | C13H12N2O | 212.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
Catalog number ACR302972500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mg
Request bulk or custom format
Specifications
Chemical Name or MaterialHarmine
Name Note98%
CAS442-51-3
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: