Scopoletin, 95%, Thermo Scientific Chemicals
Scopoletin, 95%, Thermo Scientific Chemicals
Scopoletin, 95%, Thermo Scientific Chemicals
Scopoletin, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Scopoletin, 95%, Thermo Scientific Chemicals
CAS: 92-61-5 | C10H8O4 | 192.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
250 mg
1 g
Packaging:
Glass bottle
Catalog number ACR302900500
Price (MYR)
621.00
Add to cart
Quantity:
50 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
621.00
Add to cart
Chemical Identifiers
CAS76620-00-3
IUPAC Name1-(4-ethylphenyl)-2,5-dihydro-1H-pyrrole-2,5-dione
Molecular FormulaC12H11NO2
InChI KeyFHVHFKZQDVQILM-UHFFFAOYSA-N
SMILESCCC1=CC=C(C=C1)N1C(=O)C=CC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)62.0-69.0?C
FormCrystals or powder or crystalline powder or lumps
Assay (GC)≥98.0%
Appearance (Color)Pale yellow to yellow