3,4-(Methylenedioxy)phenylacetic acid, 98.5+%, Thermo Scientific Chemicals
3,4-(Methylenedioxy)phenylacetic acid, 98.5+%, Thermo Scientific Chemicals
3,4-(Methylenedioxy)phenylacetic acid, 98.5+%, Thermo Scientific Chemicals
3,4-(Methylenedioxy)phenylacetic acid, 98.5+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3,4-(Methylenedioxy)phenylacetic acid, 98.5+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 g
Packaging:
Glass bottle
Catalog number ACR302620025
Price (MYR)
477.00
Add to cart
Quantity:
2.5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
477.00
Add to cart
Specifications
Chemical Name or Material3, 4-(Methylenedioxy)phenylacetic acid
CAS2861-28-1
CAS Min %98.5
CAS Max %100.0
Health Hazard 2GHS H Statement: