2-Chloroethylphosphonic acid, 90%, Tech., Thermo Scientific Chemicals
2-Chloroethylphosphonic acid, 90%, Tech., Thermo Scientific Chemicals
2-Chloroethylphosphonic acid, 90%, Tech., Thermo Scientific Chemicals
2-Chloroethylphosphonic acid, 90%, Tech., Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Chloroethylphosphonic acid, 90%, Tech., Thermo Scientific Chemicals
CAS: 16672-87-0 | C2H6ClO3P | 144.49 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number ACR301630050
Price (MYR)
3,284.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,284.00
Add to cart
Chemical Identifiers
CAS530-93-8
IUPAC Name1,2,3,4-tetrahydronaphthalen-2-one
Molecular FormulaC10H10O
InChI KeyKCKZIWSINLBROE-UHFFFAOYSA-N
SMILESO=C1CCC2=CC=CC=C2C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to green
Infrared spectrumConforms
Refractive index1.5550 to 1.5650 (20°C, 589 nm)
Appearance (Form)Liquid or low melting solid
GC>=94 %