Lithium tert-butoxide, 99%, Thermo Scientific Chemicals
Lithium tert-butoxide, 99%, Thermo Scientific Chemicals
Lithium tert-butoxide, 99%, Thermo Scientific Chemicals
Lithium tert-butoxide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Lithium tert-butoxide, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number ACR301220250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialLithium tert-butoxide
CAS1907-33-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Flammable solid.
Harmful if swallowed.
Reacts violently with water.
Health Hazard 3GHS P Statement
Keep cool.
Protect from sunlight.
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if