trans-2-Nonenal, 95%, Thermo Scientific™
trans-2-Nonenal, 95%, Thermo Scientific™
trans-2-Nonenal, 95%, Thermo Scientific™
trans-2-Nonenal, 95%, Thermo Scientific™
Thermo Scientific Chemicals
trans-2-Nonenal, 95%, Thermo Scientific™
Have Questions?
Catalog NumberQuantityPackaging
ACR29831025025 mLGlass bottle
Catalog number ACR298310250
Price (MYR)
916.00
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
916.00
Add to cart
Specifications
Chemical Name or Materialtrans-2-Nonenal
Name Note95%
CAS18829-56-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement