β-Ionone, 96%, synthetic, Thermo Scientific Chemicals
β-Ionone, 96%, synthetic, Thermo Scientific Chemicals
β-Ionone, 96%, synthetic, Thermo Scientific Chemicals
β-Ionone, 96%, synthetic, Thermo Scientific Chemicals
Thermo Scientific Chemicals
β-Ionone, 96%, synthetic, Thermo Scientific Chemicals
CAS: 14901-07-6 | C13H20O | 192.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number ACR297131000
Price (MYR)
595.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
595.00
Add to cart
Chemical Identifiers
CAS108-68-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale yellow to brown
FormPowder or crystals or crystalline powder or fused solid
Assay (GC)≥98.0%
Melting Point60.0-66.0°C