(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one, 98%, Thermo Scientific Chemicals
(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one, 98%, Thermo Scientific Chemicals
(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one, 98%, Thermo Scientific Chemicals
(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one, 98%, Thermo Scientific Chemicals
CAS: 49805-30-3 | C6H7NO | 109.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number ACR297060010
Price (MYR)
515.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
515.00
Add to cart
Chemical Identifiers
CAS109425-51-6
IUPAC Name2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-[1-(triphenylmethyl)-1H-imidazol-4-yl]propanoic acid
Molecular FormulaC40H33N3O4
InChI KeyXXMYDXUIZKNHDT-UHFFFAOYNA-N
SMILESOC(=O)C(CC1=CN(C=N1)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)97.5% min