5-Hexyn-1-ol, 97%, Thermo Scientific Chemicals
5-Hexyn-1-ol, 97%, Thermo Scientific Chemicals
5-Hexyn-1-ol, 97%, Thermo Scientific Chemicals
5-Hexyn-1-ol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5-Hexyn-1-ol, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number ACR296580050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7681-11-0
IUPAC Namepotassium iodide
Molecular FormulaIK
InChI KeyNLKNQRATVPKPDG-UHFFFAOYSA-M
SMILES[K+].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentBa 0.002%
Trace Metals(as Pb) 5ppm
Impurity contentIO₃ 3ppm
pHof a 5% solution 6.0-9.2 at 25?
Assay (unspecified)99.0% KI min