(Chloromethylene)dimethylammonium chloride, 96%, Thermo Scientific Chemicals
(Chloromethylene)dimethylammonium chloride, 96%, Thermo Scientific Chemicals
(Chloromethylene)dimethylammonium chloride, 96%, Thermo Scientific Chemicals
(Chloromethylene)dimethylammonium chloride, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
(Chloromethylene)dimethylammonium chloride, 96%, Thermo Scientific Chemicals
CAS: 3724-43-4 | C3H7Cl2N | 128 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number ACR295770050
Price (MYR)
521.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
521.00
Add to cart
Chemical Identifiers
CAS6153-56-6
IUPAC Nameoxalic acid dihydrate
Molecular FormulaC2H6O6
InChI KeyGEVPUGOOGXGPIO-UHFFFAOYSA-N
SMILESO.O.OC(=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Melting Point (clear melt)98-106?C
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Alkylation reagent
RUO – Research Use Only