2,2,4-Trimethylpentane, 99+%, pure, Thermo Scientific Chemicals
2,2,4-Trimethylpentane, 99+%, pure, Thermo Scientific Chemicals
2,2,4-Trimethylpentane, 99+%, pure, Thermo Scientific Chemicals
2,2,4-Trimethylpentane, 99+%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,2,4-Trimethylpentane, 99+%, pure, Thermo Scientific Chemicals
CAS: 540-84-1 | C8H18 | 114.232 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
Packaging:
Glass Bottle
Catalog number ACR295720025
Price (MYR)
711.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
711.00
Add to cart
Chemical Identifiers
CAS4949-69-3
IUPAC Name4-iodo-3-methylaniline
Molecular FormulaC7H8IN
InChI KeyUISBOJCPTKUBIC-UHFFFAOYSA-N
SMILESCC1=CC(N)=CC=C1I
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Beige to light brown to dark grey
Appearance (Form)Crystalline powder
Infrared spectrumConforms
GC>=97.5 %
Melting point50°C to 55°C