Phosphoric acid, extra pure, 85% solution in water, Thermo Scientific Chemicals
Phosphoric acid, extra pure, 85% solution in water, Thermo Scientific Chemicals
Phosphoric acid, extra pure, 85% solution in water, Thermo Scientific Chemicals
Phosphoric acid, extra pure, 85% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phosphoric acid, extra pure, 85% solution in water, Thermo Scientific Chemicals
CAS: 7664-38-2 | H3O4P | 97.994 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number ACR295700010
Price (MYR)
678.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
678.00
Add to cart
Chemical Identifiers
CAS9002-92-0
MDL NumberMFCD00043063
SpecificationsSpecification SheetSpecification Sheet
Acid Value≤1.0 mg KOH/g
Colour (Hazen Units)0-50
Appearance (Color)White
FormWaxy fused solid or pellets or flakes