Ethylene glycol, 99.5%, for analysis, Thermo Scientific Chemicals
Ethylene glycol, 99.5%, for analysis, Thermo Scientific Chemicals
Ethylene glycol, 99.5%, for analysis, Thermo Scientific Chemicals
Ethylene glycol, 99.5%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethylene glycol, 99.5%, for analysis, Thermo Scientific Chemicals
CAS: 107-21-1 | C2H6O2 | 62.068 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
1 L
2.5 L
Packaging:
View all
Metal Drum
Catalog number ACR295530250
Price (MYR)
4,792.00
Add to cart
Quantity:
25 L
Packaging:
Metal Drum
Request bulk or custom format
Price (MYR)
4,792.00
Add to cart
Chemical Identifiers
CAS22013-33-8
IUPAC Name2,3-dihydro-1,4-benzodioxin-6-amine
Molecular FormulaC8H9NO2
InChI KeyBZKOZYWGZKRTIB-UHFFFAOYSA-N
SMILESNC1=CC=C2OCCOC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
Appearance (Color)Pale yellow to yellow to dark red to dark brown to brown to black
FormCrystals or powder or crystalline powder or fused solid or clear liquid or viscous liquid as melt
Refractive Index1.5985-1.6015 @ 20?C
For analysis
RUO – Research Use Only