Ethylene glycol, 99.5%, for analysis, Thermo Scientific Chemicals
Ethylene glycol, 99.5%, for analysis, Thermo Scientific Chemicals
Ethylene glycol, 99.5%, for analysis, Thermo Scientific Chemicals
Ethylene glycol, 99.5%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethylene glycol, 99.5%, for analysis, Thermo Scientific Chemicals
CAS: 107-21-1 | C2H6O2 | 62.068 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
25 L
Packaging:
View all
Glass Bottle
Catalog number ACR295530025
Price (MYR)
794.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
794.00
Add to cart
Chemical Identifiers
CAS101-81-5
IUPAC Namebenzylbenzene
Molecular FormulaC13H12
InChI KeyCZZYITDELCSZES-UHFFFAOYSA-N
SMILESC(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Low melting solid
GC>=98.5 %
Appearance (Color)Colorless to light yellow
Infrared spectrumConforms
For analysis
RUO – Research Use Only