Ethylene glycol, 99.5%, for analysis, Thermo Scientific Chemicals
Ethylene glycol, 99.5%, for analysis, Thermo Scientific Chemicals
Ethylene glycol, 99.5%, for analysis, Thermo Scientific Chemicals
Ethylene glycol, 99.5%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethylene glycol, 99.5%, for analysis, Thermo Scientific Chemicals
CAS: 107-21-1 | C2H6O2 | 62.068 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
25 L
Packaging:
View all
Glass Bottle
Catalog number ACR295530010
Price (MYR)
606.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
606.00
Add to cart
Chemical Identifiers
CAS1314-61-0
IUPAC Name[(dioxotantalio)oxy]dioxotantalum
Molecular FormulaO5Ta2
InChI KeyPBCFLUZVCVVTBY-UHFFFAOYSA-N
SMILESO=[Ta](=O)O[Ta](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Total trace metal impurities=<0.01 %
Appearance (Form)Powder
For analysis
RUO – Research Use Only