Chromosulfuric acid, Thermo Scientific Chemicals
Chromosulfuric acid, Thermo Scientific Chemicals
Chromosulfuric acid, Thermo Scientific Chemicals
Chromosulfuric acid, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chromosulfuric acid, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass bottle
Catalog number ACR295510010
Price (MYR)
769.00
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
769.00
Add to cart
Chemical Identifiers
CAS10035-06-0
IUPAC Namebismuth(3+) pentahydrate trinitrate
Molecular FormulaBiH10N3O14
InChI KeyFBXVOTBTGXARNA-UHFFFAOYSA-N
SMILESO.O.O.O.O.[Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White or transparent
Appearance (Form)Crystalline powder or crystals and/or chunks
Titration Complexometric>=98.0 %
Insoluble matter=<0.005 %
Chloride (Cl)=<0.001 %
View more
Oxidation reagent
RUO – Research Use Only