Fullerene C60, 99.9%, Thermo Scientific Chemicals
Fullerene C60, 99.9%, Thermo Scientific Chemicals
Fullerene C60, 99.9%, Thermo Scientific Chemicals
Fullerene C60, 99.9%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Fullerene C60, 99.9%, Thermo Scientific Chemicals
CAS: 99685-96-8 | C60 | 720.66 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Catalog number ACR295012500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS56442-17-2
IUPAC Name4-[(4-fluorophenyl)methoxy]benzaldehyde
Molecular FormulaC14H11FO2
InChI KeyQGULWBQOCMQNFD-UHFFFAOYSA-N
SMILESFC1=CC=C(COC2=CC=C(C=O)C=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Free acid (titration)≤2.0%
Appearance (Color)White to pale cream
Melting Point (clear melt)95.0-101.0?C
Assay (GC)≥96.0%