Fullerene C60, 99.9%, Thermo Scientific Chemicals
Fullerene C60, 99.9%, Thermo Scientific Chemicals
Fullerene C60, 99.9%, Thermo Scientific Chemicals
Fullerene C60, 99.9%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Fullerene C60, 99.9%, Thermo Scientific Chemicals
CAS: 99685-96-8 | C60 | 720.66 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Packaging:
Glass bottle
Catalog number ACR295010010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialFullerene C60
Name Note99.9%
CAS99685-96-8
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: