7-Methoxy-2-tetralone, 95%, Thermo Scientific™
7-Methoxy-2-tetralone, 95%, Thermo Scientific™
7-Methoxy-2-tetralone, 95%, Thermo Scientific™
7-Methoxy-2-tetralone, 95%, Thermo Scientific™
Thermo Scientific Chemicals
7-Methoxy-2-tetralone, 95%, Thermo Scientific™
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number ACR294970050
Price (MYR)
615.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
615.00
Add to cart
Specifications
Chemical Name or Material7-Methoxy-2-tetralone
CAS4133-34-0
CAS Min %94.0
CAS Max %100.0
ColorYellow to Orange