3,4-Dichlorobenzaldehyde, 98%
3,4-Dichlorobenzaldehyde, 98%
3,4-Dichlorobenzaldehyde, 98%
3,4-Dichlorobenzaldehyde, 98%
Thermo Scientific Chemicals
3,4-Dichlorobenzaldehyde, 98%
CAS: 6287-38-3 | C7H4Cl2O | 175.01 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR29456025025 gGlass bottle
Catalog number ACR294560250
Price (MYR)
609.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
609.00
Add to cart
Chemical Identifiers
CAS5382-16-1
IUPAC Name4-hydroxypiperidin-1-ium
Molecular FormulaC5H12NO
InChI KeyHDOWRFHMPULYOA-UHFFFAOYSA-O
SMILESOC1CC[NH2+]CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<1 % (K.F.)
GC>=99.0 %
Appearance (Color)White to yellow
Appearance (Form)Crystalline powder or crystals and/or chunks