Bis(diphenylphosphino)methane, 98%, Thermo Scientific Chemicals
Bis(diphenylphosphino)methane, 98%, Thermo Scientific Chemicals
Bis(diphenylphosphino)methane, 98%, Thermo Scientific Chemicals
Bis(diphenylphosphino)methane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Bis(diphenylphosphino)methane, 98%, Thermo Scientific Chemicals
CAS: 2071-20-7 | C25H22P2 | 384.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number ACR293610250
Price (MYR)
1,808.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,808.00
Add to cart
Chemical Identifiers
CAS107-35-7
IUPAC Name2-aminoethane-1-sulfonic acid
Molecular FormulaC2H7NO3S
InChI KeyXOAAWQZATWQOTB-UHFFFAOYSA-N
SMILESNCCS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or crystalline powder or needles
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5% (non-U.S. specification)
Assay from Suppliers CofA≥98.5% (U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated