4-Cyanostyrene, 95%, stabilized, Thermo Scientific Chemicals
4-Cyanostyrene, 95%, stabilized, Thermo Scientific Chemicals
4-Cyanostyrene, 95%, stabilized, Thermo Scientific Chemicals
4-Cyanostyrene, 95%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Cyanostyrene, 95%, stabilized, Thermo Scientific Chemicals
CAS: 3435-51-6 | C9H7N | 129.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 g
Packaging:
Glass bottle
Catalog number ACR292910025
Price (MYR)
1,307.00
Add to cart
Quantity:
2.5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,307.00
Add to cart
Specifications
Chemical Name or Material4-Cyanostyrene
CAS3435-51-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if swallowed.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor