2-Bromoacetamide, 98%, Thermo Scientific Chemicals
2-Bromoacetamide, 98%, Thermo Scientific Chemicals
2-Bromoacetamide, 98%, Thermo Scientific Chemicals
2-Bromoacetamide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Bromoacetamide, 98%, Thermo Scientific Chemicals
CAS: 683-57-8 | C2H4BrNO | 137.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number ACR291100050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Bromoacetamide
Name Note98%
CAS683-57-8
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger