4-tert-Butylcalix[8]arene, 97%, Thermo Scientific™
4-tert-Butylcalix[8]arene, 97%, Thermo Scientific™
4-tert-Butylcalix[8]arene, 97%, Thermo Scientific™
4-tert-Butylcalix[8]arene, 97%, Thermo Scientific™
4-tert-Butylcalix[8]arene, 97%, Thermo Scientific™
Have Questions?
Catalog NumberQuantityPackaging
ACR29024025025 gGlass bottle
Catalog number ACR290240250
Price (MYR)
600.96
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
600.96
Add to cart
Specifications
Chemical Name or Material4-tert-Butylcalix[8]arene
Name Note97%
CAS68971-82-4
CAS Min %96.0
CAS Max %100.0
View more
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chelation/Complexation reagent
RUO – Research Use Only