Calix[4]arene, 98%, Thermo Scientific Chemicals
Calix[4]arene, 98%, Thermo Scientific Chemicals
Calix[4]arene, 98%, Thermo Scientific Chemicals
Calix[4]arene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Calix[4]arene, 98%, Thermo Scientific Chemicals
CAS: 74568-07-3 | C28H24O4 | 424.5 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number ACR290190050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialCalix[4]arene
Name Note98%
CAS74568-07-3
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
View more
Chelation/Complexation reagent
RUO – Research Use Only