Calix[4]arene, 98%, Thermo Scientific Chemicals
Calix[4]arene, 98%, Thermo Scientific Chemicals
Calix[4]arene, 98%, Thermo Scientific Chemicals
Calix[4]arene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Calix[4]arene, 98%, Thermo Scientific Chemicals
CAS: 74568-07-3 | C28H24O4 | 424.5 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number ACR290190010
Price (MYR)
1,146.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,146.00
Add to cart
Chemical Identifiers
CAS89-83-8
IUPAC Name5-methyl-2-(propan-2-yl)phenol
Molecular FormulaC10H14O
InChI KeyMGSRCZKZVOBKFT-UHFFFAOYSA-N
SMILESCC(C)C1=CC=C(C)C=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥98.0%
Melting Point47.5-52.5?C
Appearance (Color)White
Identification (FTIR)Conforms
Chelation/Complexation reagent
RUO – Research Use Only