Polysorbate 80, Thermo Scientific Chemicals
Polysorbate 80, Thermo Scientific Chemicals
Polysorbate 80, Thermo Scientific Chemicals
Polysorbate 80, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Polysorbate 80, Thermo Scientific Chemicals
Polysorbate 80, CAS Number-9005-65-6, Polyoxyethylene(20)sorbitan monooleate, 2.5L, 2mg KOH/g max., Yellow to Amber, 1.0800g/mL, 5ppm max., >110 deg.C, 65 to 80mg KOH/g, Authentic, 15, 7703, MFCD00082107, Glass bottle, Viscous Liquid At 25 deg.C, Indoors +20 deg.C, 1.4710 to 1.4730, 45 to 55mg KOH/g
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
10 L
Catalog number ACR278632500
Price (MYR)
425.00
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (MYR)
425.00
Add to cart
Chemical Identifiers
CAS2315-36-8
IUPAC Name2-chloro-N,N-diethylacetamide
Molecular FormulaC6H12ClNO
InChI KeyCQQUWTMMFMJEFE-UHFFFAOYSA-N
SMILESCCN(CC)C(=O)CCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to brown
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4680 to 1.4720 (20°C, 589 nm)
GC>=97.5 %
RUO – Research Use Only
Contents & Storage
Indoors +20°C