Polysorbate 80, Thermo Scientific Chemicals
Polysorbate 80, Thermo Scientific Chemicals
Polysorbate 80, Thermo Scientific Chemicals
Polysorbate 80, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Polysorbate 80, Thermo Scientific Chemicals
Polysorbate 80, CAS Number-9005-65-6, Polyoxyethylene(20)sorbitan monooleate, 2.5L, 2mg KOH/g max., Yellow to Amber, 1.0800g/mL, 5ppm max., >110 deg.C, 65 to 80mg KOH/g, Authentic, 15, 7703, MFCD00082107, Glass bottle, Viscous Liquid At 25 deg.C, Indoors +20 deg.C, 1.4710 to 1.4730, 45 to 55mg KOH/g
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
10 L
Catalog number ACR278630025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS7789-45-9
IUPAC Namecopper(2+) dibromide
Molecular FormulaBr2Cu
InChI KeyQTMDXZNDVAMKGV-UHFFFAOYSA-L
SMILES[Cu++].[Br-].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration Iodometric>=99.0 %
Sulfate (SO4)=<2000 ppm
Appearance (Color)Dark grey to grey-black
Insoluble matter=<0.05 % (in water)
View more
RUO – Research Use Only
Contents & Storage
Indoors +20°C