Polysorbate 80, Thermo Scientific Chemicals
Polysorbate 80, Thermo Scientific Chemicals
Polysorbate 80, Thermo Scientific Chemicals
Polysorbate 80, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Polysorbate 80, Thermo Scientific Chemicals
Polysorbate 80, CAS Number-9005-65-6, Polyoxyethylene(20)sorbitan monooleate, 2.5L, 2mg KOH/g max., Yellow to Amber, 1.0800g/mL, 5ppm max., >110 deg.C, 65 to 80mg KOH/g, Authentic, 15, 7703, MFCD00082107, Glass bottle, Viscous Liquid At 25 deg.C, Indoors +20 deg.C, 1.4710 to 1.4730, 45 to 55mg KOH/g
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
2.5 L
10 L
Catalog number ACR278630010
Price (MYR)
772.00
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
772.00
Add to cart
Specifications
Chemical Name or MaterialPolysorbate 80
CAS9005-65-6
ColorYellow to Amber
Density1.0800 g/mL
Quantity1 L
View more
RUO – Research Use Only
Contents & Storage
Indoors +20°C