Potassium hydroxide, 1N solution in water, Thermo Scientific Chemicals
Potassium hydroxide, 1N solution in water, Thermo Scientific Chemicals
Potassium hydroxide, 1N solution in water, Thermo Scientific Chemicals
Potassium hydroxide, 1N solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Potassium hydroxide, 1N solution in water, Thermo Scientific Chemicals
CAS: 1310-58-3 | HKO | 56.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Plastic bottle
Catalog number ACR278400025
Price (MYR)
570.00
Add to cart
Quantity:
2.5 L
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
570.00
Add to cart
Chemical Identifiers
CAS1310-58-3, 7732-18-5
Specifications
Chemical Name or MaterialPotassium hydroxide
ColorColorless
Name Note1N solution in water
Boiling Point100.0°C
CAS1310-58-3