Potassium hydroxide, 1N solution in water, Thermo Scientific Chemicals
Potassium hydroxide, 1N solution in water, Thermo Scientific Chemicals
Potassium hydroxide, 1N solution in water, Thermo Scientific Chemicals
Potassium hydroxide, 1N solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Potassium hydroxide, 1N solution in water, Thermo Scientific Chemicals
CAS: 1310-58-3 | HKO | 56.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Plastic bottle
Catalog number ACR278400010
Price (MYR)
447.00
Add to cart
Quantity:
1 L
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
447.00
Add to cart
Chemical Identifiers
CAS10034-96-5
IUPAC Namemanganese(2+) hydrate sulfate
Molecular FormulaH2MnO5S
InChI KeyISPYRSDWRDQNSW-UHFFFAOYSA-L
SMILESO.[Mn++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.0-101.0% MnSO₄?H₂O
Impurity contentCa 0.005%
Impurity contentK 0.01%
Impurity contentSubstances reducing permanganate P.T.
Insoluble material0.01%