Carbopol™ 940
Carbopol™ 940
Carbopol™ 940
Thermo Scientific Chemicals
Carbopol™ 940
CAS: 9003-01-04 | (C3H4O2)n | 72.06 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR278382500250 gPlastic bottle
Catalog number ACR278382500
Price (MYR)
1,881.00
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,881.00
Add to cart
Chemical Identifiers
CAS2390-09-2
IUPAC Namemethyl (11E)-icos-11-enoate
Molecular FormulaC21H40O2
InChI KeyRBKMRGOHCLRTLZ-ZHACJKMWSA-N
SMILESCCCCCCCC\C=C\CCCCCCCCCC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
NMR (1H-NMR)Consistent with structure
GC>=98.5 %