Petroleum ether, for analysis, boiling range 80-110°C, Thermo Scientific Chemicals
Petroleum ether, for analysis, boiling range 80-110°C, Thermo Scientific Chemicals
Petroleum ether, for analysis, boiling range 80-110°C, Thermo Scientific Chemicals
Petroleum ether, for analysis, boiling range 80-110°C, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Petroleum ether, for analysis, boiling range 80-110°C, Thermo Scientific Chemicals
CAS: 64742-49-0 | C6H14 | 86.178 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass Bottle
Catalog number ACR278230010
Price (MYR)
482.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
482.00
Add to cart
Chemical Identifiers
CAS10361-91-8
IUPAC Nameytterbium(3+) trichloride
Molecular FormulaCl3Yb
InChI KeyCKLHRQNQYIJFFX-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Yb+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
For analysis
RUO – Research Use Only