2-Aminoterephthalic acid, 99%, Thermo Scientific Chemicals
2-Aminoterephthalic acid, 99%, Thermo Scientific Chemicals
2-Aminoterephthalic acid, 99%, Thermo Scientific Chemicals
2-Aminoterephthalic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Aminoterephthalic acid, 99%, Thermo Scientific Chemicals
CAS: 10312-55-7 | C8H5NO4 | 179.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number ACR278031000
Price (MYR)
2,005.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,005.00
Add to cart
Chemical Identifiers
CAS3597-91-9
IUPAC Name{[1,1'-biphenyl]-4-yl}methanol
Molecular FormulaC13H12O
InChI KeyAXCHZLOJGKSWLV-UHFFFAOYSA-N
SMILESOCC1=CC=C(C=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥98.0% (non-U.S. specification)
Assay from Suppliers CofA≥98.0% (U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
Melting Point (clear melt)96-108°C