2-Aminoterephthalic acid, 99%, Thermo Scientific Chemicals
2-Aminoterephthalic acid, 99%, Thermo Scientific Chemicals
2-Aminoterephthalic acid, 99%, Thermo Scientific Chemicals
2-Aminoterephthalic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Aminoterephthalic acid, 99%, Thermo Scientific Chemicals
CAS: 10312-55-7 | C8H5NO4 | 179.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number ACR278030250
Price (MYR)
835.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
835.00
Add to cart
Chemical Identifiers
CAS103-81-1
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>98.5%