1H,1H-Perfluoro-1-octanol, 98%, Thermo Scientific Chemicals
1H,1H-Perfluoro-1-octanol, 98%, Thermo Scientific Chemicals
1H,1H-Perfluoro-1-octanol, 98%, Thermo Scientific Chemicals
1H,1H-Perfluoro-1-octanol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1H,1H-Perfluoro-1-octanol, 98%, Thermo Scientific Chemicals
CAS: 307-30-2 | C8H3F15O | 400.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number ACR277260050
Price (MYR)
622.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
622.00
Add to cart
Chemical Identifiers
CAS1312-81-8
IUPAC Namedilanthanum(3+) trioxidandiide
Molecular FormulaLa2O3
InChI KeyMRELNEQAGSRDBK-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[La+3].[La+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Total Rare Earth Oxide Impurities≤0.001% (10 ppm)