5-Methoxy-2-mercaptobenzimidazole, 99+%, Thermo Scientific Chemicals
5-Methoxy-2-mercaptobenzimidazole, 99+%, Thermo Scientific Chemicals
5-Methoxy-2-mercaptobenzimidazole, 99+%, Thermo Scientific Chemicals
5-Methoxy-2-mercaptobenzimidazole, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5-Methoxy-2-mercaptobenzimidazole, 99+%, Thermo Scientific Chemicals
CAS: 37052-78-1 | C8H8N2OS | 180.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number ACR275370250
Price (MYR)
1,673.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,673.00
Add to cart
Specifications
Chemical Name or Material5-Methoxy-2-mercaptobenzimidazole
Name Note99+%
CAS37052-78-1
Absorbance(1% soln. in methanol) (1 cm cell) no other absorption observed from 360 to 600nm, (1% soln. in methanol) (1 cm cell) no other absorption observed from 360 to 600nm at 0nm, 0.4 max. at 360nm
CAS Min %99.0