5-Methoxy-2-mercaptobenzimidazole, 99+%, Thermo Scientific Chemicals
5-Methoxy-2-mercaptobenzimidazole, 99+%, Thermo Scientific Chemicals
5-Methoxy-2-mercaptobenzimidazole, 99+%, Thermo Scientific Chemicals
5-Methoxy-2-mercaptobenzimidazole, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5-Methoxy-2-mercaptobenzimidazole, 99+%, Thermo Scientific Chemicals
CAS: 37052-78-1 | C8H8N2OS | 180.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number ACR275370050
Price (MYR)
566.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
566.00
Add to cart
Chemical Identifiers
CAS9002-88-4
Molecular Formula(C2H4)n
SMILES*-CC-*
MDL NumberMFCD00084423
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Particle Sizemean particle size 150 micron