Paraldehyde, 99.5%, Thermo Scientific Chemicals
Product Image
Thermo Scientific Chemicals
Paraldehyde, 99.5%, Thermo Scientific Chemicals
CAS: 123-63-7 | C6H12O3 | 132.16 g/mol
Have Questions?
Catalog NumberQuantity
ACR2750800101 L
Catalog number ACR275080010
Price (MYR)
703.00
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
703.00
Add to cart
Chemical Identifiers
CAS95-47-6
IUPAC Name1,2-xylene
Molecular FormulaC8H10
InChI KeyCTQNGGLPUBDAKN-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.5035 to 1.5065 (20°C, 589 nm)
GC>=98.5 %
Residue after evaporation=<0.01 %